Bovine serum albumin (BSA)
Leverandør: ENZO LIFE SCIENCES
ADI801928
ENZOADI801928EA 1100 NOK
ENZOADI801928
Bovine serum albumin (BSA)
Proteiner
This BSA Solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in ImmunoSet products. Each bottle is sufficient for use with 5×96 wells microtiter plates if following the instructions in our ImmunoSet protocol. The BSA Solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our ImmunoSet products.
New Product
Bestill nå
SPESIFIKASJONER
Protein/peptide name Bovine Serum Albumin
Environmentally Preferable
Protein synonyms BSA|Fraction V|Cohn Fraction V
Arter Bovine
Kilde Serum
Konsentrasjon 10%