Urinary prostacyclin ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900025
ENZOADI900025EA 6475 NOK
ENZOADI900025
Urinary prostacyclin ELISA kit
Assay ELISAs
For the quantitative determination of urinary prostacyclin metabolites in culture supernatants and urine from any species.
The Urinary Prostacyclin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 by measuring the stable metabolites in one simple assay.
Bestillingsinformasjon: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-292 (5x96 wells)