Gastrin I (human), ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900026
ENZOADI900026EA 6875 NOK
ENZOADI900026
Gastrin I (human), ELISA kit
Assay ELISAs
For the quantitative determination of human Gastrin I in culture supernatants, plasma, and serum.
The Gastrin I (human), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405nm. Ready-to-use liquid color-coded reagents are provided to reduce error in the lab.
Bestillingsinformasjon: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.