IL-4 (mouse), ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900043
ENZOADI900043EA 5225 NOK
ENZOADI900043
IL-4 (mouse), ELISA kit
Assay ELISAs
For the quantitative determination of mouse IL-4 in culture supernatants and serum. Cited sample type includes plasma.
The IL-4 (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.
Bestillingsinformasjon: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.