cAMP ELISA Kits
Leverandør: ENZO LIFE SCIENCES
ADI900067A
ENZOADI900067AEA 6050 NOK
ENZOADI900067A
cAMP ELISA Kits
Assay ELISAs
Most sensitive (0,039pmol/m) colorimetric kit for extracellular cAMP quantification.
For the quantitative determination of cAMP in culture supernatants, serum, and saliva from any species. Cited sample types include cell lysate, tissue, and urine.
This is a colorimetric competitive enzyme immunoassay kit for the quantitative determination of extracellular cAMP. Sensitivity is increased 10-fold by acetylation (reagents included). Absorbance is read at 405 nm. The ready-to-use liquid color-coded reagents provided reduce error.
Bestillingsinformasjon: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 2, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine. Alternative size available: 89141-312 (5x96 wells).