LTB4 ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900068
ENZOADI900068EA 6125 NOK
ENZOADI900068
LTB4 ELISA kit
Assay ELISAs
For the quantitative determination of LTB4 in culture supernatants, plasma, saliva, and urine from any species. Cited sample types include bronchioalveolar lavage fluid, cell lysate, crevicular fluid, tissue, serum and whole blood.
The LTB4 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Assay up to 39 samples in duplicate with a broad dynamic range that is suitable for a wide variety of sample matrices. Document your results to scientists world-wide with this highly cited kit.
Bestillingsinformasjon: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-314 (5x96 wells)