PGF2α ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900069
ENZOADI900069EA 5950 NOK
ENZOADI900069
PGF2α ELISA kit
Assay ELISAs
For the quantitative determination of PGF2α in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include cell lysate and whole blood.
The PGF2α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Broad dynamic range is suitable for a wide variety of sample types. Ready-to-use liquid color-coded reagents are safe, non-radioactive, and reduce laboratory errors.
Bestillingsinformasjon: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-316 (5x96 wells)