Cysteinyl leukotriene ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900070
ENZOADI900070EA 6475 NOK
ENZOADI900070
Cysteinyl leukotriene ELISA kit
Assay ELISAs
For the quantitative determination of Cysteinyl Leukotrienes (LTC4/D4/E4) in culture supernatants and urine from any species.
The Cysteinyl Leukotriene EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Safe non-radioactive measurement of LTC4/D4/E4. Ready-to-use liquid color-coded reagents reduce errors.
Bestillingsinformasjon: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-318 (5x96 wells)