Big endothelin-1 (rat), ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900073
ENZOADI900073EA 13125 NOK
ENZOADI900073
Big endothelin-1 (rat), ELISA kit
Assay ELISAs
The Big Endothelin-1 (Rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results overnight + 1 hour.
  • Measure low levels of rat big endothelin-1
  • Convenient method
  • Assay up to 38 samples in duplicate per plate
For the quantitative determination of rat Big ET-1 in culture supernatants, plasma, and serum.
Leveringsinformasjon: Kit contains microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.