DHEA ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900093
ENZOADI900093EA 5975 NOK
ENZOADI900093
DHEA ELISA kit
Assay ELISAs
Highly sensitive ELISA, measuring DHEA in culture supernatants, plasma, serum, saliva, and urine from any species.
For the quantitative determination of DHEA in culture supernatants, plasma, serum, saliva, and urine from any species.
The DHEA EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit has low reactivity to structurally related molecules and offers sensitive measurement of DHEA in a variety of sample types.
Bestillingsinformasjon: Contains GxR IgG Microtiter plate, Assay buffer, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-3263 (5x96 wells).