COX-2 (human), ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900094
ENZOADI900094EA 13125 NOK
ENZOADI900094
COX-2 (human), ELISA kit
Assay ELISAs
The COX-2 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours. This is the most sensitive COX-II immunoassay kit commercially available for inflammation research.
  • Easy to use
  • lThe color-coded reagents reduce errors
  • Reliable and fast
For the quantitative determination of human COX-2 in cell lysates.
Leveringsinformasjon: Supplied with microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.