[pThr180/Tyr182]p38 ELISA Kit
Leverandør: ENZO LIFE SCIENCES
ADI900101
ENZOADI900101EA 10550 NOK
ENZOADI900101
[pThr180/Tyr182]p38 ELISA Kit
Assay ELISAs
For the quantitative determination of [pThr180/Tyr182]p38 in cell lysates of human and mouse origin.
The [pThr180/Tyr182]p38 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. Use less precious sample per test compared to Western blot with higher throughput sample analysis; measure up to 40 samples in duplicate per kit.
Bestillingsinformasjon: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 4 concentrate, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.