Osteopontin (human), ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900142
ENZOADI900142EA 8850 NOK
ENZOADI900142
Osteopontin (human), ELISA kit
Assay ELISAs
For the quantitative determination of human Osteopontin in culture supernatants, milk, plasma, and urine. Cited sample type includes serum.
The Osteopontin (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. Ready-to-use liquid color-coded reagents reduce error. Excellent publications in peer-reviewed publications demonstrate the utility and consistency of this kit developed with highly cited double monoclonal antibodies.
Bestillingsinformasjon: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.