IGF-1 (human), ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900150
ENZOADI900150EA 9225 NOK
ENZOADI900150
IGF-1 (human), ELISA kit
Assay ELISAs
Highly sensitive ELISA, measuring as little as 50pg/ml of IGF-1 in just 4 hours.
For the quantitative determination of human IGF-1 in plasma and serum.
The IGF-1 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450 nm. This kit offers ultra-sensitive measurement of as little as 50pg/ml of IGF-1.
Bestillingsinformasjon: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Standard, Wash buffer concentrate, TMB substrate, Stop solution 2, Neutralizing reagent 2, 2N HCl.