IL-17A (human), ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900177
ENZOADI900177EA 8475 NOK
ENZOADI900177
IL-17A (human), ELISA kit
Assay ELISAs
A highly sensitive competitive immunoassay for the measurement of IL-17A in human culture supernatants, serum and plasma samples. Results from up to 40 duplicate samples are read on a colorimetric plate reader at 450nm in just 3 hours.
  • Ultra-Sensitive
  • Negligible reactivity with other family members
  • Pre-coated plates and liquid color-coded reagents reduce errors
For the quantitative determination of human IL-17A in culture supernatants, plasma, and serum samples.
Leveringsinformasjon: Supplied with microtiter plate, standard, antibody, conjugate, assay buffer 13, TMB substrate, stop solution 2, wash buffer concentrate.