Bradykinin ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI900206
ENZOADI900206EA 7675 NOK
ENZOADI900206
Bradykinin ELISA kit
Assay ELISAs
The Bradykinin ELISA kit is a colorimetric, competitive immunoassay kit for the quantitative determination of species independent Bradykinin in plasma, serum and urine samples.
  • Sensitive - measure as little as 24,8 pg/ml of Bradykinin
  • Rapid results in just 3 hours from up to 41 samples in duplicate
  • Broad dynamic range to easily quantify high and low levels of Bradykinin
  • Easy to use liquid colour coded reagents reduce errors
For the quantitative determination of species independent Bradykinin in plasma, serum and urine samples.
The Bradykinin EIA kit is a colorimetric, competitive immunoassay kit with results in 3 hours.
Bestillingsinformasjon: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.
Leveringsinformasjon: The kit contains microtiter plate, conjugate, antibody, assay buffer 16, wash buffer concentrate, standard, TMB substrate, stop solution 2, and Streptavidin-HRP.