Survival Motor Neuron (SMN) ELISA Kits
Leverandør: ENZO LIFE SCIENCES
ADI900209
ENZOADI900209EA 9375 NOK
ENZOADI900209
Survival Motor Neuron (SMN) ELISA Kits
Assay ELISAs
The SMN ELISA Kit is a colorimetric immunometric immunoassay kit for the quantitative determination of human and mouse SMN in cell lysate samples.
  • Highly sensitive measurement, detecting as little as 50 pg/ml of SMN
  • High throughput format with results in <3 hours for up to 39 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
  • Convenient - ready to use liquid colour coded reagents
  • Pre-coated plate saves time and minimise errors
Leveringsinformasjon: The kit contains microtiter plate, conjugate, antibody, assay buffer 13, wash buffer concentrate, standard, TMB substrate, stop solution 2, and extraction reagent 4.