24(S)-Hydroxycholesterol ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI9002100001
ENZOADI9002100001EA 8900 NOK
ENZOADI9002100001
24(S)-Hydroxycholesterol ELISA kit
Assay ELISAs
Only quantitative 24(S)-hydroxycholesterol ELISA kit available for neurodegenerative research.
For the quantitative determination of species independent 24(S)-Hydroxycholesterol in serum, plasma, cerebral spinal fluid, culture supernatants and tissue samples.
The 24(S)-Hydroxycholesterol ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of 24(S)-Hydroxycholesterol in serum, plasma, tissue culture media (culture supernatant), cerebral spinal fluid and tissue homogenate samples with results in just 2 hours.
Bestillingsinformasjon: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.