25(OH) Vitamin D ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI9002150001
ENZOADI9002150001EA 10425 NOK
ENZOADI9002150001
25(OH) Vitamin D ELISA kit
Assay ELISAs
Rapid, easy-to-use ELISA kit to quantify 25(OH) Vitamin D for bone metabolism, nutrition, endocrinology, immunity and cancer.
For the quantitative determination of human 25(OH) Vitamin D in plasma and serum.
The 25(OH) Vitamin D ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of 25(OH) Vitamin D in plasma and serum with results in just 1.5 hours.
Bestillingsinformasjon: Contains Microtiter Plate, Conjugate Concentrate, Antibody, Dissociation Buffer, Conjugate Diluent, Sample Diluent, Wash Buffer Concentrate, Standards, Substrate, Stop Solution, Assay Layout.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Mål 25-(OH) Vitamin D
Primary antibody reactivity
Analysetype Competitive
Vert
Sensitivitet 1,98 ng/ml (range 0,5 - 1010 ng/ml)