5-Hydroxymethylcytosine DNA ELISA Kit, Grow Cells
Leverandør: ENZO LIFE SCIENCES
ADI9002250001
ENZOADI9002250001EA 8375 NOK
ENZOADI9002250001
5-Hydroxymethylcytosine DNA ELISA Kit, Grow Cells
Assay ELISAs
Highly sensitive, rapid ELISA kit to quantify 5-hydroxymethylcytosine in DNA with results that closely correlate to mass spectrometry analysis.
  • Rapid: Fully quantitative results in less than three hours
  • Convenient: User-friendly alternative to mass spectrometry
  • Simple: Efficient and well-established protocol
  • High-throughput: 96-well format ideal for global detection, tissue-specific quantitation, high-throughput compound screening, and more
For the quantitative determination of species independent 5-hydroxymethylcytosine DNA in a variety of DNA samples.
The 5-Hydroxymethylcytosine DNA ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of 5-hydroxymethylcytosine in DNA  with results in just three hours.
Bestillingsinformasjon: Contains Microtiter Plate, Coating Buffer, ELISA Buffer, Primary Antibody, Antibody-conjugate,  HRP Developer, Controls.