Urinary prostacyclin ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI901025
ENZOADI901025EA 25875 NOK
ENZOADI901025
Urinary prostacyclin ELISA kit
Assay ELISAs
For the quantitative determination of urinary prostacyclin metabolites in culture supernatants and urine from any species.
The Urinary Prostacyclin ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 by measuring the stable metabolites in one simple assay.
Bestillingsinformasjon: Contains DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-084 (1x96 wells).