12(S)-HETE ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI901050
ENZOADI901050EA 29725 NOK
ENZOADI901050
12(S)-HETE ELISA kit
Assay ELISAs
For the quantitative determination of 12(S)-HETE in culture supernatants and plasma from any species. Cited sample types include serum, renal interstitial fluid and tissue.
The 12(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. Fast, accurate, and reliable results are obtained with ready-to-use liquid color-coded reagents that reduce error.
Bestillingsinformasjon: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-102 (1x96 wells).