15(S)-HETE ELISA kit
Leverandør: ENZO LIFE SCIENCES
ADI901051
ENZOADI901051EA 24225 NOK
ENZOADI901051
15(S)-HETE ELISA kit
Assay ELISAs
For the quantitative determination of 15(S)-HETE in culture supernatants, plasma, serum, and urine from any species.
The 15(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.
Bestillingsinformasjon: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-104 (1x96 wells).