sPLA2 activity kit
Leverandør: ENZO LIFE SCIENCES
ADI907002
ENZOADI907002EA 7275 NOK
ENZOADI907002
sPLA2 activity kit
Assay Cellulær analyse
For the quantitative determination of sPLA2 in culture supernatants, cell lysates, and buffers from any species.
  • Higher throughput - assay up to 42 samples in duplicate per 96 well kit
  • Sensitive - detect < 4 U/ml
  • Fast - results in just 40 minutes
The sPLA2 activity kit is a higher throughput assay for the quantification of up to 42 samples in duplicate per 96 well kit. This kit provides safe, sensitive, and reliable results in under an hour.
Bestillingsinformasjon: Contains Microtiter plate, Standard, Reaction buffer concentrate, Substrate, Stop reagent, Color reagent.