Myeloperoxidase fluorometric detection kit
Leverandør: ENZO LIFE SCIENCES
ADI907029
ENZOADI907029EA 10825 NOK
ENZOADI907029
Myeloperoxidase fluorometric detection kit
Assay Cellulær analyse
The Myeloperoxidase fluorometric detection kit is a one-step no wash homogeneous activity assay that is adaptable to HTS applications. This kit offers highly sensitive fluorescent detection and the ability to monitor multiple time points to follow the kinetics of MPO activity.
  • Fast homogenous assay
  • Suitable for High Throughput Screening
  • Monitor multiple time points to follow kinetics
  • Results in 30 to 60 minutes
For the measurement of Myeloperoxidase activity in cells and tissue from any species.
Leveringsinformasjon: Assay buffer concentrate, Detection Reagent, Hydrogen peroxide, Myeloperoxidase enzyme
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Sensitivitet 0,00195 units/ml
Størrelse 500 tests
Lagringsforhold 4 °C (long term)