Creatinine colorimetric detection kit
Leverandør: ENZO LIFE SCIENCES
ADI907030A
ENZOADI907030AEA 3550 NOK
ENZOADI907030A
Creatinine colorimetric detection kit
Kolorimeter Komparatorsett
The creatinine colorimetric detection kit utilises a single step liquid detection reagent that is safer and less time consuming than other assay methods.
  • Easy-to-use one step creatinine detection
  • Fast homogeneous mix-and-ready assay with no wash steps, results in just 30 minutes
  • Accurate and calibrated to NIST creatinine standard
This kit is calibrated against the NIST standard and offers reproducible results with less than 6% inter- and intra-assay variation.