PGE2 CLIA kit
Leverandør: ENZO LIFE SCIENCES
ADI910001
ENZOADI910001EA 6950 NOK
ENZOADI910001
PGE2 CLIA kit
Assay ELISAs
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species.
The PGE2 CLIA kit is a sensitive chemiluminescent competitive enzyme immunoassay kit for the quantitative determination of PGE2 in biological fluids from any species.
Bestillingsinformasjon: Contains White GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, CLIA Substrate.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Mål PGE2
Primary antibody reactivity
Analysetype Competitive
Vert
Sensitivitet 6,03 pg/ml (range 7,81 - 1,000 pg/ml)