Nitric Oxide (NO2-/NO3-) detection kit
Leverandør: ENZO LIFE SCIENCES
ADI917010
ENZOADI917010EA 4325 NOK
ENZOADI917010
Nitric Oxide (NO2-/NO3-) detection kit
Assay Cellulær analyse
For the quantitative determination of nitrite and nitrate levels in culture supernatants, plasma, serum, saliva, and urine from any species.
The Nitric Oxide (NO2-/NO3-), detection kit is a complete kit for the quantitative determination of nitrate and nitrite concentrations. Absorbance is read at 540-570 nm. Only 0.050 mL diluted sample needed per well saves precious sample.
Bestillingsinformasjon: Contains Microtiter plate, Nitrate Reductase, Nitrate Reductase Storage Buffer, NADH, Nitrate Standard, Nitrite Standard, Reaction buffer concentrate, Griess Reagent I, Griess Reagent II.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Sensitivitet (Nitrite) 0,222 µmole/L; (Nitrate) 0,625 µmole/L (range (Nitrite) 3,125 - 200 µmole/L; (Nitrate) 3,125 - 100 µmole/L)