Nitric Oxide (NO2-/NO3-) detection kit
Leverandør: ENZO LIFE SCIENCES
ADI917010
ENZOADI917010EA 4325 NOK
ENZOADI917010
Nitric Oxide (NO2-/NO3-) detection kit
Assay Cellulær analyse
For the quantitative determination of nitrite and nitrate levels in culture supernatants, plasma, serum, saliva, and urine from any species.
The Nitric Oxide (NO2-/NO3-), detection kit is a complete kit for the quantitative determination of nitrate and nitrite concentrations. Absorbance is read at 540-570 nm. Only 0.050 mL diluted sample needed per well saves precious sample.
Bestillingsinformasjon: Contains Microtiter plate, Nitrate Reductase, Nitrate Reductase Storage Buffer, NADH, Nitrate Standard, Nitrite Standard, Reaction buffer concentrate, Griess Reagent I, Griess Reagent II.