Mouse-HRP/rabbit-AP IHC, MULTIVIEW®
Leverandør: ENZO LIFE SCIENCES
ADI9501000001
ENZOADI9501000001EA 6825 NOK
ENZOADI9501000001
Mouse-HRP/rabbit-AP IHC, MULTIVIEW®
Assay Immunologisk analyser
Mouse-HRP/rabbit-AP IHC is a biotin-free one-step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and cryostat sections.
  • Nanopolymer-based system for superior multiplex antigen detection
  • Expandable with additional chromogens
  • Preserves precious tissue while reducing reagents
May be used with blood smears, cytosmears, and cell preparations.
Bestillingsinformasjon: Includes a peroxidase block, POLYVIEW® IHC reagent (mouse-HRP), POLYVIEW® IHC reagent (rabbit-AP), HIGHDEF® IHC chromogen substrate (DAB, stable) buffer and chromogen, HIGHDEF® red IHC chromogen (AP) buffer and chromogen, empty mixing bottles.