Yellow IHC chromogen (HRP), HighDef®
Leverandør: ENZO LIFE SCIENCES
ADI9501700030
ENZOADI9501700030EA 2925 NOK
ENZOADI9501700030
Yellow IHC chromogen (HRP), HighDef®
Assay Cellulær analyse
Yellow IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridisation systems.
  • High definition yellow IHC chromogen for HRP-activated development
  • Distinct bright yellow that can easily be distinguished from other stains
  • This system can be used in any automation system as well as manual use
Leveringsinformasjon: Substrate buffer and chromogen in an empty mixing bottle.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringsforhold Long term
Lagringstemp 4 °C