Black IHC chromogen (HRP), HIGHDEF®
Leverandør: ENZO LIFE SCIENCES
ADI9501710030
ENZOADI9501710030EA 2875 NOK
ENZOADI9501710030
Black IHC chromogen (HRP), HIGHDEF®
Assay Cellulær analyse
HIGHDEF® black IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridisation systems.
  • Unique high definition black IHC chromogen for HRP-activated development
  • System used in any automation system as well as manual use
  • Produces a sharply contrasting ebony black colour
Leveringsinformasjon: Black IHC chromogen (HRP) substrate buffer and chromogen in an empty mixing bottle.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringsforhold Long term
Lagringstemp 4 °C