IHC enhancer (DAB, plus), HIGHDEF®
Leverandør: ENZO LIFE SCIENCES
ADI9501820015
ENZOADI9501820015EA 2425 NOK
ENZOADI9501820015
IHC enhancer (DAB, plus), HIGHDEF®
Assay Immunologisk analyser
HIGHDEF® IHC enhancer (DAB, plus) is a DAB signal enhancer used to increase intensity of immunostaining using DAB as chromogen.
  • Superior performance compared to amino ethyl carbazol (AEC)
HIGHDEF® IHC enhancer (DAB, plus) increases the signal intensity without creating any background staining.