IHC chromogen substrate (AEC, stable), HIGHDEF®
Leverandør: ENZO LIFE SCIENCES
ADI9502100030
ENZOADI9502100030EA 2050 NOK
ENZOADI9502100030
IHC chromogen substrate (AEC, stable), HIGHDEF®
Elektroforese reagenser Western Blotting Reagents
HIGHDEF® red IHC chromogen can be used in conjunction with peroxidase-based immunostaining systems.
Peroxidase from the immunodetection system reacts with hydrogen peroxide substrate to degrade it, which reacts with AEC precipitating it at positive sites yielding a reddish brown product.