IHC universal negative control reagent
Leverandør: ENZO LIFE SCIENCES
ADI9502310025
ENZOADI9502310025EA 1925 NOK
ENZOADI9502310025
IHC universal negative control reagent
Assay Immunologisk analyser
IHC universal negative control reagent is specifically designed to work with rabbit, mouse and goat antibodies.
  • Liquid mixture of purified goat, mouse and rabbit immunoglobulins (IgGs)
The reagent has been titered to work with polymer-based secondary systems in manual and automated IHC platforms.