Vacuum manifold
Leverandør: ENZO LIFE SCIENCES
ADI987500
ENZOADI987500EA 5975 NOK
ENZOADI987500
Vacuum manifold
Manifold
The manifold is constructed of a chemical-resistant polymer and is designed to facilitate the filtration of liquid through a multi-well filter plate.