Anti-Bcl-xL Mouse Monoclonal Antibody [clone: 2H12]
Leverandør: ENZO LIFE SCIENCES
ADIAAM080E
ENZOADIAAM080EEA 5675 NOK
ENZOADIAAM080E
Anti-Bcl-xL Mouse Monoclonal Antibody [clone: 2H12]
Antistoffer
The Bcl-2 family of proteins regulate the intrinsic apoptotic pathway and are characterized by the sharing of at least one of four conserved Bcl-2 Homology (BH) domains.
  • Dilution Range: 1-5 µg/ml, ECL for Western blot
  • Detects a band of ~28kDa by Western blot
Bcl-2 family members function as either pro- or anti-apoptotic factors, and are generally categorized into three groups based on structural homology and function. Bcl-2-like proteins (Bcl-2, Bcl-xL, Bcl-w, A1, and Mcl-1) are pro-survival. Pro-apoptotic members can be sub-divided into two groups:More Product Information
Bestill nå
SPESIFIKASJONER
Antigensymbol BCL2L1
Environmentally Preferable
Antigen-navn BCL2-like 1
Antigen synonymer BCLX|BCL2L1|Apoptosis regulator Bcl-X|BLC2L|BCL2L|Bcl-2-like 1 protein
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon 2H12
Reaktivitet
Vert Mouse
Isotype IgG2a
Western blot Yes
Epitop near the N-Terminus
Immunogen Synthetic peptide corresponding to the sequence near the N-terminus of human Bcl-xL
Format Ascites
Skjema Liquid
Lagringsbuffer PBS containing 0,08% sodium azide.
Lagringstemp –20 °C
Frakttemp. Shipped on blue ice