Anti-Caspase-10 Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
ADIAAP110E
ENZOADIAAP110EEA 5625 NOK
ENZOADIAAP110E
Anti-Caspase-10 Rabbit Polyclonal Antibody
Antistoffer
Caspases are synthesized as inactive zymogens that can be cleaved to form active enzymes following the induction of apoptosis by stress or death receptors.
  • Dilution Range: 2 µg/ml, ECL for Western blot
  • Detects a band of ~59 kDa by Western blot
Initiator caspases are activated by dimerization of the zymogen on a dedicated adaptor protein. These activated initiator caspases in-turn cleave downstream effector or executioner caspases in a cascade-like manner, which cleave key cellular proteins that lead to the morphological changes associated withMore Product Information
Bestill nå
SPESIFIKASJONER
Antigensymbol CASP10
Environmentally Preferable
Antigen-navn Caspase 10
Antigen synonymer Caspase 10 apoptosis-related cysteine peptidase|FLICE2|ALPS2|MCH4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype
Western blot Yes
Immunogen Synthetic peptide corresponding to aa 505-521 of human Caspase-10
Aminosyretall 505 to 521
Skjema Liquid
Rensing Purified by DEAE
Lagringsbuffer PBS containing 0,02% sodium azide
Lagringstemp 4 °C
Frakttemp. Blue ice not frozen