Anti-CASP9 Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
ADIAAP149E
ENZOADIAAP149EEA 4550 NOK
ENZOADIAAP149E
Anti-CASP9 Rabbit Polyclonal Antibody
Antistoffer
The caspases are a family of cysteine proteases that cleave after certain aspartate residues, and are primarily recognized as mediators of apoptosis. caspases are synthesized as inactive zymogens that can be cleaved to form active enzymes following the induction of apoptosis by stress or death receptors. Initiator caspases (e.g. caspase-8 and -10) are activated by dimerization of the zymogen on a dedicated adaptor protein. These activated initiator caspases in-turn cleave downstream effector or executioner caspases (e.g. caspase-3, -6, and -7) in a cascade-like manner, which cleave keyMore Product Information