Anti-MD-2 Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ADICSA506E
ENZOADICSA506EEA 5875 NOK
ENZOADICSA506E
Anti-MD-2 Rabbit Polyclonal Antibody
Antistoffer
MD-2 is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs recognize different pathogen-associated molecular patterns, thereby linking the innate and adaptive immune responses.

  • Dilution range: 2 µg/ml for Immunohistochemistry, 0,5 to 2,0 µg/ml for Western blot

TLR4, the major signaling receptor for lipopolysaccharide (LPS), requires the binding of MD-2 to its extracellular region for maximal response to LPS.

Type: Primary
Antigen: Lymphocyte antigen 96
Clonality: Polyclonal
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol LY96
Environmentally Preferable
Antigen-navn Lymphocyte antigen 96
Antigen synonymer ESOP-1|MD2|MD-2|ly-96|lymphocyte antigen 96
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Isotype
Western blot Yes
ImmunoKjemi Yes
Immunogen Synthetic peptide corresponding to the sequence near the middle of human MD-2
Skjema Liquid
Rensing Purified by ion exchange chromatography
Lagringsbuffer PBS containing 0,02% sodium azide
Lagringstemp 4 °C
Frakttemp. blue ice not frozen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR