Anti-ST2 Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ADICSA512E
ENZOADICSA512EEA 5625 NOK
ENZOADICSA512E
Anti-ST2 Rabbit Polyclonal Antibody
Antistoffer
ST2 is a member of a superfamily containing the interleukin-1 receptor and the Toll-like receptors (TLRs).

  • Dilution Range: 2 µg/ml for Immunohistochemistry, 1-2 µg/ml for Western blot
  • Detects a band of ~45kDa by Western blot

ST2 was originally identified as a protein whose production was stimulated by various proliferation inducing agents such as PDGF and FGF. Additionally, it has been shown to negatively regulate IL-1 receptor and Toll-like receptor (TLR) 4 signaling and to maintain endotoxin tolerance.

Type: Primary
Antigen: Interleukin 1 Receptor-More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR