DNA damage ELISA Kit
Leverandør: ENZO LIFE SCIENCES
ADIEKS350
ENZOADIEKS350EA 12775 NOK
ENZOADIEKS350
DNA damage ELISA Kit
Assay ELISAs
Rapid DNA damage ELISA kit for cancer, apoptosis and oxidative stress.
For the measurement of DNA Damage (8-OHdG) in serum, saliva, and urine from any species. Cited sample types include plasma, seminal fluids, cell lysates, culture supernatants and DNA extracts.
The DNA Damage EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 2,5 hours. This kit has been validated in-house in a variety of sample matrices and is supplied in a convenient 96 well plate format. Protocol (xls) available upon request.
Bestillingsinformasjon: Contains Microtiter plate, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Primary antibody reactivity
Analysetype Competitive
Vert
Sensitivitet 0,59 ng/ml (range 1,875 - 60 ng/ml)