PKA kinase activity kit
Leverandør: ENZO LIFE SCIENCES
ADIEKS390A
ENZOADIEKS390AEA 8825 NOK
ENZOADIEKS390A
PKA kinase activity kit
Assay Enzymanalyser
The PKA Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the quantitative determination of PKA kinase activity in partially purified, purified, or crude enzyme preparations from any species.
Bestillingsinformasjon: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2