PKC kinase activity kit
Leverandør: ENZO LIFE SCIENCES
ADIEKS420A
ENZOADIEKS420AEA 8675 NOK
ENZOADIEKS420A
PKC kinase activity kit
Assay Enzymanalyser
The PKC Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the measurement of PKC kinase activity partially purified, purified, or crude enzyme preparations from any species. Cited sample type includes tissue (ref. 1).
Bestillingsinformasjon: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2