p70 S6K activity kit
Leverandør: ENZO LIFE SCIENCES
ADIEKS470
ENZOADIEKS470EA 10325 NOK
ENZOADIEKS470
p70 S6K activity kit
Assay Cellulær analyse
The p70 S6K activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the measurement of p70 S6K activity in partially purified, purified, or crude enzyme preparations from any species.
Bestillingsinformasjon: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2