HSP70 high sensitivity ELISA Kit
Leverandør: ENZO LIFE SCIENCES
ADIEKS715
ENZOADIEKS715EA 9600 NOK
ENZOADIEKS715
HSP70 high sensitivity ELISA Kit
Assay ELISAs
Only commercially available kit specifically validated and sensitive enough to quantify HSP70 (HSP72) in serum and plasma samples.
For the quantitative determination of HSP70 in serum and plasma of human, mouse, and rat origin. Cited sample(s) include cell lysates and urine.
The HSP70 high sensitivity EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4,5 hours. Absorbance is read at 450 nm. This is the only commercially available kit specifically validated and sensitive enough to quantify HSP70 (HSP72) in serum and plasma samples, as referenced by the Cell Stress Society International.
Bestillingsinformasjon: Contains Microtiter plate, Assay buffer 28, Standard, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2.