Anti-AcK Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
ADIKAPTF120E
ENZOADIKAPTF120EEA 6450 NOK
ENZOADIKAPTF120E
Anti-AcK Rabbit Polyclonal Antibody
Antistoffer
Acetylation and methylation of lysine are important post-translational modifications that regulate numerous protein-protein and protein-DNA interactions. Lysine acetylation and methylation involves the transfer of acetylCoA, or one or more methyl groups, to the e-amino group of lysine by modifying enzymes and cofactors. Histones and transcription factors are the primary targets of lysine acetylation and methylation, with either modification capable of inducing gene silencing or expression due to differential regulation of cofactors. For example, varying degrees of mono-, di-, and tri-More Product Information