Anti-HMOX1 Mouse Monoclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ADIOSA111D ADIOSA111F ADIOSA111J
ENZOADIOSA111DEA 4175 NOK
ENZOADIOSA111D ENZOADIOSA111F ENZOADIOSA111J
Anti-HMOX1 Mouse Monoclonal Antibody
Antistoffer
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin. These products of the HO reaction have important physiological effects: carbon monoxide is a potent vasodilator and has been implicated to be a physiological regulator of cGMP and vascular tone; biliverdin and its product bilirubin are potent antioxidants; "free" iron increases oxidative stress and regulates the expression ofMore Product Information
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol HMOX1 HMOX1
Environmentally Preferable
Antigen-navn Heme Oxygenase 1 Heme Oxygenase 1
Antigen synonymer Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1 Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
Antistofftype Primary Primary
Klonalitet Monoclonal Monoclonal
Reaktivitet Human
Vert Mouse Mouse
Isotype IgG2b IgG2b
Western blot Yes Yes
ImmunoKjemi Yes Yes
Flowcytometri Yes Yes
Skjema Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide. Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
Rensing Protein G affinity purified Protein G affinity purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR