Anti-HMOX1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: ENZO LIFE SCIENCES
ADIOSA111FID ADIOSA111FIF
ENZOADIOSA111FIDEA 3575 NOK
ENZOADIOSA111FID ENZOADIOSA111FIF
Anti-HMOX1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin. These products of the HO reaction have important physiological effects: carbon monoxide is a potent vasodilator and has been implicated to be a physiological regulator of cGMP and vascular tone; biliverdin and its product bilirubin are potent antioxidants; "free" iron increases oxidative stress and regulates the expression ofMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol HMOX1 HMOX1
Environmentally Preferable
Antigen-navn Heme Oxygenase 1 Heme Oxygenase 1
Antigen synonymer Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1 Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
Antistofftype Primary Primary
Klonalitet Monoclonal Monoclonal
Konjugering FITC (Fluorescein Isothiocyanate) FITC (Fluorescein Isothiocyanate)
Reaktivitet Human
Vert Mouse Mouse
Isotype IgG2b IgG2b
Flowcytometri Yes Yes
Skjema Liquid. In PBS containing 0,09% sodium azide. Liquid. In PBS containing 0,09% sodium azide.
Rensing Protein G affinity purified Protein G affinity purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR