Anti-HMOX1 Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ADIOSA150F
ENZOADIOSA150FEA 6675 NOK
ENZOADIOSA150F
Anti-HMOX1 Rabbit Polyclonal Antibody
Antistoffer
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin.

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR