Anti-GRPE Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
ADISPA240J
ENZOADISPA240JEA 24825 NOK
ENZOADISPA240J
Anti-GRPE Rabbit Polyclonal Antibody
Antistoffer
GrpE (Glucose Regulated Protein E. coli) is encoded by a monocistronic E. coli gene, which is under the control of both s32 and s70 transcription factors. The GrpE protein is a so-called co-chaperone because it is known to assist the DnaK (Hsp70) protein to effectively carry out DnaK-dependent chaperone activity (i.e. protein folding, protein transport, disaggregation of heat inactivated proteins, activation of mutant protein).

Type: Primary
Antigen: GrpE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR